missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Tubulin Antibody (YL1/2) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 38 publications
£147.00 - £432.00
Specifications
| Antigen | alpha Tubulin |
|---|---|
| Clone | YL1/2 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:5000-1:10000, Flow Cytometry 2-5 ug/0.1x10^6 cells, ELISA 1:100-1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:1000-1:10000. Use reported in scientific literature (PMID 28001364), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:200, Functional reported in scientific literature (PMID 31358662), Radioimmunoassay |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Radioimmune Assays (RIA) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18717013
|
Novus Biologicals
NB600-506SS |
0.025 mL |
£147.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18795683
|
Novus Biologicals
NB600-506 |
0.1 mL |
£432.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha Tubulin Monoclonal specifically detects alpha Tubulin in Human, Mouse, Rat, Avian, C. elegans, Drosophila, Invertebrate, Mammal, Monkey, Primate, Yeast samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Functional, Radioimmunoassay.Specifications
| alpha Tubulin | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Functional Assay, Radioimmune Assays (RIA) | |
| Unconjugated | |
| Rat | |
| Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Loading Controls, Neuroscience | |
| Alpha-tubulin 3, B-ALPHA-1, FLJ25113, hum-a-tub1, hum-a-tub2, LIS3, TUBA3, tubulin alpha-1A chain, Tubulin alpha-3 chain, Tubulin B-alpha-1, tubulin, alpha 1a, tubulin, alpha 3, tubulin, alpha, brain-specific | |
| TUBA1A | |
| IgG2a | |
| Protein G purified | |
| Human, mouse, rat, monkey, S. cerevisiae, S. pombe, Slime moulds, Allium and birds. Expected to react with most eukaryotes due to sequence identity. Drosophila reactivity reported in scientific literature (PMID: 24019759) Other species have not been tested. |
| YL1/2 | |
| Western Blot 1:5000-1:10000, Flow Cytometry 2-5 ug/0.1x10^6 cells, ELISA 1:100-1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:1000-1:10000. Use reported in scientific literature (PMID 28001364), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen 1:200, Functional reported in scientific literature (PMID 31358662), Radioimmunoassay | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Avian, C. elegans, Drosophila, Invertebrate, Mammalia, Monkey, Primate, Yeast | |
| 7846 | |
| This alpha Tubulin Antibody (YL1/2) was developed against full length native protein (purified) (S. cerevisiae). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 50 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title