missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladin Antibody (1E6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-25959CL1
This item is not returnable.
View return policy
Description
Palladin Monoclonal antibody specifically detects Palladin in Canine,Chicken,Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/Immunofluorescence
Specifications
| Palladin | |
| Monoclonal | |
| CoraFluor 1 | |
| CGI151, CGI-151, FLJ22190, FLJ38193, KIAA0992FLJ39139, MYN, myoneurin, palladin, palladin, cytoskeletal associated protein, PNCA1, Sarcoma antigen NY-SAR-77, SIH002FLJ61376 | |
| Endogenous Palladin immunoprecipitated from chicken embryo fibroblasts. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Signal Transduction | |
| 23022 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 1E6 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Canine, Chicken, Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction