missing translation for 'onlineSavingsMsg'
Learn More

Paliperidone Palmitate, Mikromol™

Product Code. 30051018
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051018 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051018 Supplier Mikromol Supplier No. MM3627.000250
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Paliperidone Palmitate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Paliperidone Palmitate
CAS 199739-10-1
Analyte or Component Names Paliperidone Palmitate
Shipping Condition Room Temperature
Molecular Formula C39 H57 F N4 O4
Certifications/Compliance ISO 17034
Synonym Hexadecanoic acid, 3-[2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl ester, 9-Hydroxyrisperidone palmitate, Invega Sustenna, Paliperidone palmitate, RO 92670
InChI Formula InChI=1S/C39H57FN4O4/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-18-36(45)47-34-17-16-24-44-38(34)41-29(2)32(39(44)46)23-27-43-25-21-30(22-26-43)37-33-20-19-31(40)28-35(33)48-42-37/h19-20,28,30,34H,3-18,21-27H2,1-2H3
SMILES CCCCCCCCCCCCCCCC(=O)OC1CCCN2C(=O)C(=C(C)N=C12)CCN3CCC(CC3)c4noc5cc(F)ccc45
IUPAC Name [3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-4-oxo-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-9-yl] hexadecanoate
Molecular Weight (g/mol) 664.89
Quantity 250 mg
Formula Weight 664.4364 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.