missing translation for 'onlineSavingsMsg'
Learn More

Paliperidone, MedChemExpress

Product Code. p-200100030
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16497986

Brand: Medchem Express HYA0019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Paliperidone (9-Hydroxyrisperidone), the major active metabolite of Risperidone, is a dopamine D2 antagonist and 5-HT2A antagonist. Paliperidone is also active as an antagonist at α1 and α2 adrenergic receptors and H1-histaminergic receptors. Paliperidone, a antipsychotic agent, shows efficacy against schizophrenia.

Paliperidone (9-Hydroxyrisperidone), the major active metabolite of Risperidone, is a dopamine D2 antagonist and 5-HT2A antagonist. Paliperidone is also active as an antagonist at α1 and α2 adrenergic receptors and H1-histaminergic receptors. Paliperidone, a antipsychotic agent, shows efficacy against schizophrenia.

Specifications

Chemical Name or Material Paliperidone
CAS 144598-75-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H27FN4O3
Quantity 100 mg
Synonym 9-Hydroxyrisperidone
Solubility Information DMSO : 5 mg/mL (11.72 mM; Need ultrasonic)
SMILES CC1=C(C(N2CCCC(C2=N1)O)=O)CCN3CCC(CC3)C4=NOC5=C4C=CC(F)=C5
Molecular Weight (g/mol) 426.48
Formula Weight 426.48
Percent Purity 99.09%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.