missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palbociclib monohydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
Description
Palbociclib (PD 0332991) monohydrochloride is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. Palbociclib monohydrochloride has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, which can be used in the research of HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.
Specifications
Specifications
| Chemical Name or Material | Palbociclib monohydrochloride |
| CAS | 827022-32-2 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C24H30ClN7O2 |
| Quantity | 200 mg |
| Synonym | PD 0332991 monohydrochloride |
| Solubility Information | H2O : 7.14 mg/mL (14.75 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 4.63 mg/mL (9.57 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | O=C1C(C(C)=O)=C(C)C2=CN=C(NC3=NC=C(N4CCNCC4)C=C3)N=C2N1C5CCCC5.Cl |
| Molecular Weight (g/mol) | 483.99 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction