missing translation for 'onlineSavingsMsg'
Learn More

Palbociclib, MedChemExpress

Product Code. 16420568
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16420568

Brand: Medchem Express HY50767

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Palbociclib (PD 0332991) is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. Palbociclib has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, which can be used in the research of HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.

Palbociclib (PD 0332991) is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. Palbociclib has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, which can be used in the research of HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.

Specifications

Chemical Name or Material Palbociclib
CAS 571190-30-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H29N7O2
Quantity 5 mg
Synonym PD 0332991
Solubility Information DMSO : 25 mg/mL (55.86 mM; ultrasonic and warming and adjust pH to 5 with HCl and heat to 60°C) ∣0.1 M HCL : 25 mg/mL (55.86 mM; ultrasonic and adjust pH to 4 with 0.1 M HCL) ∣H2O : 0.1 mg/mL (0.22 mM; Need ultrasonic)
SMILES O=C1C(C(C)=O)=C(C2=CN=C(N=C2N1C3CCCC3)NC4=CC=C(N5CCNCC5)C=N4)C
Molecular Weight (g/mol) 447.53
Formula Weight 447.53
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.