missing translation for 'onlineSavingsMsg'
Learn More

PALB2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-76317-0.025mg

 View more versions of this product

Product Code. 18522431

  • £141.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PALB2 Polyclonal specifically detects PALB2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PALB2
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
DKFZp667I166, DKFZp686E1054, FANCNFLJ21816, partner and localizer of BRCA2, PNCA3
Rabbit
140 kDa
0.025 mg
Breast Cancer, Cancer
79728
Human
Purified
Western Blot
1 mg/mL
Western Blot 1-2 μg/mL, ELISA 1:100-1:2000
Q86YC2
PALB2
Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human PALB2. The immunogen is located within amino acids 940 - 990 of PALB2. Amino Acid Squence: SDDESEKQVLLKSGN
Peptide affinity purified
RUO
Primary
At least four isoforms of PALB2 are known to exist; this antibody will detect all four isoforms.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only