missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PALB2 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76317AF405
This item is not returnable.
View return policy
Description
PALB2 Polyclonal antibody specifically detects PALB2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| PALB2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 405 | |
| DKFZp667I166, DKFZp686E1054, FANCNFLJ21816, partner and localizer of BRCA2, PNCA3 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human PALB2. The immunogen is located within amino acids 940 - 990 of PALB2. Amino Acid Squence: SDDESEKQVLLKSGN | |
| 0.1 mL | |
| Breast Cancer, Cancer | |
| 79728 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction