missing translation for 'onlineSavingsMsg'
Learn More

PAL International™ PAL TX Surface Disinfectant Wipe Bucket

Product Code. 15936160
Change view
Click to view available options
Dimensions (L x W):
195 x 200 mm
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
15936160 195 x 200 mm Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15936160 Supplier PAL International™ Supplier No. W220230A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

6.0 L bucket with 1000 alcohol disinfectant wipes

  • 6.0 L Medium sized bucket of Pal TX Alcohol Disinfectant wipes
  • This handy bucket is designed for disinfecting large surfaces in high use food areas
  • Ideal for machinery and equipment, the medium bucket is fast acting and can be used to clean and disinfect food contact surfaces without leaving any residue

Specifications

Dimensions (L x W) 195 x 200 mm
Includes 6.0 L bucket with 1000 alcohol disinfectant wipes
compliance-icons
Product Identifier
  • Pal TX Alcohol Surface Disinfectant Wipes
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: propan-2-ol, ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.