missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK7 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77237AF594
This item is not returnable.
View return policy
Description
PAK7 Polyclonal antibody specifically detects PAK7 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| PAK7 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 594 | |
| PAK5 | |
| Antibody was raised against a 13 amino acid synthetic peptide from near the center of human PAK5. The immunogen is located within amino acids 240 - 290 of PAK5. Amino Acid Squence: GPSLDDYDRRPKS | |
| 0.1 mL | |
| Breast Cancer, Cancer, Protein Kinase | |
| 57144 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction