missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PAK6 Polyclonal antibody specifically detects PAK6 in Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Conjugate | Janelia Fluor 549 |
| Dilution | Western Blot |
| Gene Alias | EC 2.7.11, p21 protein (Cdc42/Rac)-activated kinase 6, p21(CDKN1A)-activated kinase 6, p21-activated kinase 6, p21-activated protein kinase 6, PAK-5, PAK5EC 2.7.11.1, PAK-6, serine/threonine-protein kinase PAK 6 |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PAK6 |
| Content And Storage | Store at 4C in the dark. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?