missing translation for 'onlineSavingsMsg'
Learn More

PAK6 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18412867 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412867 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412867 Supplier Novus Biologicals Supplier No. NBP176632V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PAK6 Polyclonal antibody specifically detects PAK6 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAK6
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, p21 protein (Cdc42/Rac)-activated kinase 6, p21(CDKN1A)-activated kinase 6, p21-activated kinase 6, p21-activated protein kinase 6, PAK-5, PAK5EC 2.7.11.1, PAK-6, serine/threonine-protein kinase PAK 6
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide from near the center of human PAK6. The immunogen is located within amino acids 260 - 310 of PAK6. Amino Acid Squence: PSSSKPGPPPQSK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Repair, GPCR, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 56924
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.