missing translation for 'onlineSavingsMsg'
Learn More

PAK4 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18410913 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410913 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410913 Supplier Novus Biologicals Supplier No. NBP177236G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PAK4 Polyclonal antibody specifically detects PAK4 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAK4
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, KIAA1142, p21 protein (Cdc42/Rac)-activated kinase 4, p21(CDKN1A)-activated kinase 4, p21-activated kinase 4, PAK-4, protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs, serine/threonine-protein kinase PAK 4
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide from near the center of human PAK4. The immunogen is located within amino acids 210 - 260 of PAK4. Amino Acid Squence: GPSAGGLAIPQS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 10298
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.