missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK3 Antibody (4G8F11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61766-0.025ml
This item is not returnable.
View return policy
Description
PAK3 Monoclonal specifically detects PAK3 in Human, Monkey samples. It is validated for Western Blot, ELISA.
Specifications
| PAK3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| beta-PAK, bPAK, CDKN1A, EC 2.7.11, EC 2.7.11.1, hPAK3, mental retardation, X-linked 47, MRX30, MRX47, Oligophrenin-3, OPHN3, p21 (CDKN1A)-activated kinase 3, p21 protein (Cdc42/Rac)-activated kinase 3, p21-activated kinase 3, PAK-3, PAK3beta, serine/threonine-protein kinase PAK 3 | |
| Mouse | |
| 62.3 kDa | |
| 0.025 mL | |
| Primary | |
| Human, Monkey | |
| Purified |
| Western Blot, ELISA | |
| 4G8F11 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PAK3 | |
| Purified recombinant fragment of human PAK3 (AA: 1-100) expressed in E. Coli. | |
| Protein G purified | |
| Protein Kinase | |
| 5063 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction