missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAK2 Recombinant Rabbit Monoclonal Antibody (23GB2595), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553292
This item is not returnable.
View return policy
Description
PAK2 (serine/threonine-protein kinase PAK2) is a serine/threonine kinase that plays a role in a variety of different signaling pathways including cytoskeleton regulation, cell motility, cell cycle progression, apoptosis, or proliferation. It acts as a downstream effector of the small GTPases CDC42 and RAC1. Activation by the binding of active CDC42 and RAC1 results in a conformation change and a subsequent autophosphorylation on several serine and/or threonine residues. Full-length PAK2 stimulates cell survival and cell growth. PAK proteins are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, serve as targets for the small GTP binding proteins Cdc42 and RAC and have been implicated in a wide range of biological activities.
Specifications
| PAK2 | |
| Recombinant Monoclonal | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13177, Q64303, Q8CIN4 | |
| Pak2 | |
| A synthesized peptide derived from human PAK2 (1-39AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2595 | |
| Unconjugated | |
| Pak2 | |
| 5330420P17Rik; A130002K10Rik; AI836325; Alpha-PAK; Beta-PAK; C-t-PAK2; D16Ertd269e; gamma pak; gamma-PAK; mKIAA4182; Oligophrenin-3; OPHN3; p21 (CDKN1A)-activated kinase 2; p21 (RAC1) activated kinase 2; p21 protein (Cdc42/Rac)-activated kinase 2; p21-activated kinase 2; p27; p34; p58; p65-PAK; PAK alpha; PAK beta; PAK gamma; PAK1; PAK-1; Pak2; PAK-2; PAK-2p27; PAK-2p34; PAK3; PAK-3; PAK65; PAKgamma; S6/H4 kinase; serine/threonine-protein kinase PAK 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 224105, 29432, 5062 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction