missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK2 Antibody (6D12), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | PAK2 |
|---|---|
| Clone | 6D12 |
| Dilution | Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunoprecipitation 1:200-1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30030357
|
Novus Biologicals
NBP3-26442-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028705
|
Novus Biologicals
NBP3-26442-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PAK2 Monoclonal antibody specifically detects PAK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunoprecipitationSpecifications
| PAK2 | |
| Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunoprecipitation 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 2.7.11, gamma-PAK, p21 (CDKN1A)-activated kinase 2, p21 protein (Cdc42/Rac)-activated kinase 2, p21-activated kinase 2, p58, PAK-2, PAK65EC 2.7.11.1, PAKgamma, S6/H4 kinase, serine/threonine-protein kinase PAK 2 | |
| A synthesized peptide derived from Human PAK2 [UniProt Q13177] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 6D12 | |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Phospho Specific, Protein Kinase | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 5062 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title