missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK2 Antibody (1E1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00005062-M01
This item is not returnable.
View return policy
Description
PAK2 Monoclonal antibody specifically detects PAK2 in Human samples. It is validated for Western Blot, ELISA, Proximity Ligation Assay, ELISA
Specifications
| PAK2 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| EC 2.7.11, gamma-PAK, p21 (CDKN1A)-activated kinase 2, p21 protein (Cdc42/Rac)-activated kinase 2, p21-activated kinase 2, p58, PAK-2, PAK65EC 2.7.11.1, PAKgamma, S6/H4 kinase, serine/threonine-protein kinase PAK 2 | |
| PAK2 (NP_002568, 131 a.a. ~ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DSNTVKQKYLSFTPPEKDGFPSGTPALNAKGTEAPAVVTEEEDDDEETAPPVIAPRPDHTKSIYTRSVIDPVPAPVGDSHVDGAAKSLDKQKKKTKMTDE | |
| 0.1 mg | |
| Protein Kinase | |
| 5062 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Proximity Ligation Assay, Sandwich ELISA | |
| 1E1 | |
| Western Blot 1:500, ELISA, Proximity Ligation Assay, Sandwich ELISA | |
| NP_002568 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction