missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAK1IP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100854
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PAK1IP1.
Negatively regulates the PAK1 kinase. PAK1 is a member of the PAK kinase family, which has been shown to play a positive role in the regulation of signaling pathways involving MAPK8 and RELA. PAK1 exists as an inactive homodimer, which is activated by binding of small GTPases such as CDC42 to an N-terminal regulatory domain. PAK1IP1 also binds to the N-terminus of PAK1, and inhibits the specific activation of PAK1 by CDC42. May be involved in ribosomal large subunit assembly (PubMed:24120868). [UniProt].
Specifications
| PAK1IP1 | |
| Polyclonal | |
| Unconjugated | |
| PAK1IP1 | |
| 5830431I15Rik; 5930415H02Rik; AA419825; AI314040; AW556169; bA421M1.5; Gdpd1; glycerophosphodiester phosphodiesterase domain containing 1; hPIP1; MAK11; p21-activated protein kinase-interacting protein 1; PAK/PLC-interacting protein 1; PAK1 interacting protein 1; PAK1-interacting protein 1; PAK1IP1; PIP1; putative PAK inhibitor Skb15; RP11-421M1.5; WD repeat-containing protein 84; WDR84 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55003, 68083 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9DCE5, Q9NWT1 | |
| PAK1IP1 | |
| A synthesized peptide derived from human PAK1IP1(Accession Q9NWT1), corresponding to amino acid residues E325-M375. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction