missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PAK1 Polyclonal antibody specifically detects PAK1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PAK1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | alpha-PAK, EC 2.7.11, EC 2.7.11.1, MGC130000, MGC130001, p21 protein (Cdc42/Rac)-activated kinase 1, p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast), p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related), p21-activated kinase 1, p65-PAK, PAK-1, PAKalpha, serine/threonine-protein kinase PAK 1, STE20 homolog, yeast |
| Host Species | Rabbit |
| Immunogen | Amino Acids encompassing and including phosphorylated Ser199 and Ser204 of human PAK1. Sequence is highly homologous (greater than 90%) to PAK2 and PAK3. |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?