missing translation for 'onlineSavingsMsg'
Learn More

PAK1 interacting protein 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-35778-100ul

 View more versions of this product

Product Code. 30229564

  • £417.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PAK1 interacting protein 1 Polyclonal antibody specifically detects PAK1 interacting protein 1 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PAK1 interacting protein 1
Polyclonal
Western Blot 1:500 - 1:2000, ELISA
bA421M1.5, FLJ20624, hPIP1WD repeat-containing protein 84, MAK11, p21-activated protein kinase-interacting protein 1, PAK1 interacting protein 1, PIP1PAK/PLC-interacting protein 1, RP11-421M1.5, WDR84PAK1-interacting protein 1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-392 of human PAK1 interacting protein 1 (NP_060376.2).,, Sequence:, MELVAGCYEQVLFGFAVHPEPEACGDHEQWTLVADFTHHAHTASLSAVAVNSRFVVTGSKDETIHIYDMKKKIEHGALVHHSGTITCLKFYGNRHLISGAEDGLICIWDAKKWECLKSIKAHKGQVTFLSIHPSGKLALSVGTDKTLRTWNLVEGRSAFIKNIKQNAHIVEWSPRGEQYVVIIQNKIDIYQLDTASISGTITNEKRISSVKFLSESVLAVAGDEEVIRFFDCDSLVCLCEFKAHENRVKDMFSFEIPEHHVIVSASSDGFIKMWKLKQDKKVPPSLLCEINTNARLTCLGVWLDKVADMKESLPPAAEPSPVSKEQSKIGKKEPGDTVHKEEKRSKPNTKKRGLTGDSKKATKESGLISTKKRKMVEMLEKKRKKKKIKTMQ
100 μL
Primary
Human, Mouse
Purified
ELISA, Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
55003
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.