missing translation for 'onlineSavingsMsg'
Learn More

PAK1 interacting protein 1 Antibody, CoraFluor™ 1, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NB100-77292CL1

Product Code. 30156656

  • £771.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PAK1 interacting protein 1 Polyclonal antibody specifically detects PAK1 interacting protein 1 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PAK1 interacting protein 1
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
55003
Store at 4°C in the dark. Do not freeze.
IgG
Immunoprecipitation, Western Blot
CoraFluor 1
bA421M1.5, FLJ20624, hPIP1WD repeat-containing protein 84, MAK11, p21-activated protein kinase-interacting protein 1, PAK1 interacting protein 1, PIP1PAK/PLC-interacting protein 1, RP11-421M1.5, WDR84PAK1-interacting protein 1
The immunogen recognized by this antibody maps to a region between residue 290 and 340 of human PAK/PLC-interacting protein 1 (WD repeat protein 84) using the numbering given in entry NP_060376.2 (GeneID 55003).
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.