missing translation for 'onlineSavingsMsg'
Learn More

PAK1 Antibody (4F10), Novus Biologicals™

Product Code. 18371475 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371475 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371475 Supplier Novus Biologicals Supplier No. NBP315141

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PAK1 Monoclonal antibody specifically detects PAK1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAK1
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 4F10
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS (pH 7.4), 150mM NaCl, and 50% glycerol.
Gene Alias alpha-PAK, EC 2.7.11, EC 2.7.11.1, MGC130000, MGC130001, p21 protein (Cdc42/Rac)-activated kinase 1, p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast), p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related), p21-activated kinase 1, p65-PAK, PAK-1, PAKalpha, serine/threonine-protein kinase PAK 1, STE20 homolog, yeast
Host Species Rabbit
Immunogen A synthesized peptide derived from human PAK1. UniProt ID (Q13153).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5058
Target Species Human, Mouse
Content And Storage Store at -20 to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.