missing translation for 'onlineSavingsMsg'
Learn More

PAIP2 Antibody (1C12), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18407845 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407845 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407845 Supplier Novus Biologicals Supplier No. NBP222555JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PAIP2 Monoclonal antibody specifically detects PAIP2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAIP2
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1C12
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias MGC72018, PABC1-interacting protein 2, PABP-interacting protein 2, PAIP-2, PAIP2APoly(A)-binding protein-interacting protein 2, poly(A) binding protein interacting protein 2, polyA-binding protein-interacting protein 2, polyadenylate-binding protein-interacting protein 2
Host Species Mouse
Immunogen Recombinant human PAIP2 (1-127aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51247
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.