missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAIP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38222-100ul
This item is not returnable.
View return policy
Description
PAIP1 Polyclonal antibody specifically detects PAIP1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| PAIP1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| MGC12360, PABC1-interacting protein 1, PABP-interacting protein 1, PAIP-1, poly(A) binding protein interacting protein 1, Poly(A)-binding protein-interacting protein 1, polyadenylate binding protein-interacting protein 1, polyadenylate-binding protein-interacting protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human PAIP1 (NP_877590.1).,, Sequence:, MSDGFDRAPEQTRPLRAPPSSQDKIPQQNSESAMAKPQVVVAPVLMSKLSVNAPEFYPSGYSSSYTESYEDGCEDYPTLSEYVQDFLNHLTEQPGSFETEIEQFAETLNGCVTTDDALQELVELIYQQATSIPNFSYMGARLCNYLSHHLTISPQSGNFRQLLLQRCRTEYEVKDQAAKGDEVTRKRFHAFVLFLGELYLNLEIKGTNGQVTRADILQVGLRELLNALFSNPMDDNLICAVKLLKLTGSVLEDAWKEKGK | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10605 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction