missing translation for 'onlineSavingsMsg'
Learn More

Thirteen Components PAH Standard solution 500μg/mL of each, Fisher Chemical™

Product Code. 12926043 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 12926043

Brand: Fisher Chemical F113981

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Chemical Name or Material 13 Components PAH Standard Solution
Packaging Ampoule
Analyte or Component Names Acenaphthylene (CAS:208-96-8), Anthracene (CAS:120-12-7), Benzo(a)anthracene (CAS:56-55-3), Benzo(j)fluoranthene (CAS:205-82-3), Benzo(k)fluoranthene (CAS:207-08-9), Benzo (g,h,i)perylene (CAS:191-24-2), Benzo(a)pyrene (CAS:50-32-8), Chrysene (CAS:218-01-9), Dibenzo (a,h)anthracene (CAS:53-70-3), Fluorene (CAS:86-73-7), Indeno(1,2,3-c,d)pyrene (CAS:193-39-5), Phenanthrene (CAS:85-01-8), Pyrene (CAS:129-00-0)
Concentration 500 μg/mL
UN Number 1294
Type Organic Multi-component Standard Solution
Element Acenaphthylene, Anthracene, Benzo(a)anthracene, Benzo(j)fluoranthene, Benzo(k)fluoranthene, Benzo (g,h,i)perylene, Benzo(a)pyrene, Chrysene, Dibenzo (a,h)anthracene, Fluorene, Indeno(1,2,3-c,d)pyrene, Phenanthrene, Pyrene
Solvent or Matrix Toluene
Quantity 1 mL
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Standard solution 13 Components in toluene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.