missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PAH Recombinant Rabbit Monoclonal Antibody (K01_1K84)

Product Code. 30829946
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829946 100 μL
30825127 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829946 Supplier Invitrogen™ Supplier No. MA563137
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

PAH Recombinant Monoclonal Antibody for Western Blot

PAH encodes the enzyme phenylalanine hydroxylase that is the rate-limiting step in phenylalanine catabolism. Deficiency of this enzyme activity results in the autosomal recessive disorder phenylketonuria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAH
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1K84
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene PAH
Gene Accession No. P00439, P04176, P16331
Gene Alias AW106920; Pah; PH; phe-4-monooxygenase; phenylalanine 4-monooxygenase; phenylalanine hydroxylase; phenylalanine-4-hydroxylase; PKU; PKU1
Gene Symbols PAH
Host Species Rabbit
Immunogen A synthetic peptide of human PAH.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18478, 24616, 5053
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.