missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119335
This item is not returnable.
View return policy
Description
PAH Polyclonal Antibody for Western Blot
PAH encodes the enzyme phenylalanine hydroxylase that is the rate-limiting step in phenylalanine catabolism. Deficiency of this enzyme activity results in the autosomal recessive disorder phenylketonuria.
Specifications
| PAH | |
| Polyclonal | |
| Unconjugated | |
| PAH | |
| AW106920; Pah; PH; phe-4-monooxygenase; phenylalanine 4-monooxygenase; phenylalanine hydroxylase; phenylalanine-4-hydroxylase; PKU; PKU1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100521900, 18478, 5053 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P00439, P16331 | |
| PAH | |
| Recombinant protein Phenylalanine Hydroxylase. The antigen corresponds to amino acid range 1-226 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction