missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PAH Monoclonal Antibody (C4), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA550896

 View more versions of this product

Product Code. 17970083

  • £161.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PAH encodes the enzyme phenylalanine hydroxylase that is the rate-limiting step in phenylalanine catabolism. Deficiency of this enzyme activity results in the autosomal recessive disorder phenylketonuria.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PAH
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
P00439, P04176, P16331
PAH
Recombinant Human PAH protein, Glu228-Lys452 (Accession #P00439).
20 μL
Primary
Human, Mouse, Pig, Rat
Antibody
IgG1 κ
Immunohistochemistry (Paraffin), Western Blot
C4
Unconjugated
PAH
AW106920; Pah; PH; phe-4-monooxygenase; phenylalanine 4-monooxygenase; phenylalanine hydroxylase; phenylalanine-4-hydroxylase; PKU; PKU1
Mouse
Protein A/G
RUO
100521900, 18478, 24616, 5053
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.