missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAH Mixture 635 0.2 μg/mL in Hexane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| CAS | 83-32-9, 208-96-8, 120-12-7, 56-55-3, 207-08-9, 50-32-8 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO/IEC 17025 |
| Solvent or Matrix | Hexane |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction