missing translation for 'onlineSavingsMsg'
Learn More

PAH Mixture 163 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30313708
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
Product Code. Quantity unitSize
30313708 5 x 1 mL 5mL
1 options
This item is not returnable. View return policy

Product Code. 30313708

Brand: Dr. Ehrenstorfer DREGS09000163DI

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

CAS 90-12-0, 91-57-6, 83-32-9, 208-96-8, 260-94-6, 120-12-7
Quantity 5 x 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.