missing translation for 'onlineSavingsMsg'
Learn More

PAH Mix A, SPEX CertiPrep™

Product Code. 15225846 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15225846 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15225846 Supplier SPEX Certiprep Supplier No. 8100A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Acenaphthene83-32-92000μg/mL
Acenaphthylene208-96-82000μg/mL
Anthracene120-12-72000μg/mL
Benz(a)anthracene56-55-32000μg/mL
Benzo(a)pyrene50-32-82000μg/mL
Benzo(b)fluoranthene205-99-22000μg/mL
Benzo(g,h,i)perylene191-24-22000μg/mL
Benzo(k)fluoranthene207-08-92000μg/mL
Carbazole86-74-82000μg/mL
Chrysene218-01-92000μg/mL
Dibenz(a,h)anthracene53-70-32000μg/mL
Fluoranthene206-44-02000μg/mL
Fluorene86-73-72000μg/mL
Indeno(1,2,3-c,d)pyrene193-39-52000μg/mL
Naphthalene91-20-32000μg/mL
Phenanthrene85-01-82000μg/mL
Pyrene129-00-02000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material PAH Mix A
Packaging 2mL amber prescored glass ampule
CAS 83-32-9, 208-96-8, 120-12-7, 56-55-3, 50-32-8, 205-99-2
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Analyte or Component Names Acenaphthene,Acenaphthylene,Anthracene
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Element Acenaphthene, Acenaphthylene, Anthracene
Solvent or Matrix MeCl:Benzene
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2000μg/mL
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • PAH MIX A
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
  • Acute toxicity Category 3
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 1A
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H350-May cause cancer.
  • H360-May damage fertility or the unborn child.
  • H372-Causes damage to organs.
  • H304-May be fatal if swallowed and enters airways.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H340-May cause genetic defects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P262-Do not get in eyes, on skin, or on clothing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P331-Do NOT induce vomiting.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: dichloromethane, benzene, anthracene, carbazole, naphthalene, chrysene

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.