missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PAGR1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12828952
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12828952 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12828952 Supplier Invitrogen™ Supplier No. PA520828

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is EL4 cell lysate. PA5-20828 can be used with blocking peptide PEP-0942.

Steroid receptor co-activators (SRCs) were initially described as nuclear receptor transcription co-activators, but they have recently been determined to co-regulate transcription initiated by other transcription factors. GAS is a recently identified glutamate-rich protein that interacts with SRC1, but not GRIP1 or AIB1, the other two members of the SRC family. GAS can also interact with the alpha subunit of the estrogen receptor (ERalpha), but not other receptors such as the retinoic acid receptor a, suggesting the interaction between GAS and ERalpha is relatively specific. Depletion of GAS by RNA interference in MCF7 cells led to a decrease in the mRNA and protein levels of ER target genes such as pS2, c-Myc and cyclin D1, indicating the role of GAS in the regulation of ER target genes. GAS has also been found to associate with an SET1-like methyltransferase complex specific for H3K4 methylation, suggesting that GAS has multiple roles in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAGR1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PAGR1
Gene Accession No. Q99L02, Q9BTK6
Gene Alias 2900092E17Rik; C16orf53; C77040; GAS; glutamate-rich coactivator associated with SRC1; glutamate-rich coactivator interacting with SRC1; glutamate-rich coactivator interacting with SRC1/NCOA1; PA1; Pagr; PAGR1; Pagr1a; PAXIP1 associated glutamate rich protein 1; PAXIP1 associated glutamate rich protein 1A; PAXIP1 associated glutamate-rich protein 1; PAXIP1-associated glutamate-rich protein 1; PAXIP1-associated glutamate-rich protein 1A; PAXIP1-associated protein 1; PTIP-associated 1; PTIP-associated 1 protein; PTIP-associated protein 1
Gene Symbols PAGR1, Pagr1a
Host Species Rabbit
Immunogen A 15 amino acid peptide from near the carboxy terminus of human GAS.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67278, 79447
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.