missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PageRuler™ Unstained Protein Ladder

Product Code. 17176098
Change view
Click to view available options
Quantity:
2 x 250 μL
8 x 250 μL
Unit Size:
Pack of 2
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17176098 8 x 250 μL Pack of 4
11802124 2 x 250 μL Pack of 2
15523197 8 x 250 μL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17176098 Supplier Thermo Scientific™ Supplier No. 26614X4
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Unstained mixture of 14 recombinant, highly purified proteins (10kDa to 200kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting.

Thermo Scientific PageRuler Unstained Protein Ladder is a mixture of 14 recombinant, highly purified, unstained proteins (10 to 200 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Features of the PageRuler Unstained Protein Ladder include:

  • Reference bands—50 kDa band is stronger than the others for easy orientation
  • Tagged—the ladder proteins contains an integral Strep-tag™ II sequence and can be detected on western blots using Strep-Tactin™ conjugates or an antibody against Strep-tag™ II Sequence

Applications

  • Accurate protein sizing on SDS-polyacrylamide gels and western blots
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contents: eight vials of 250 μL each, proteins at 0.02 to 0.05 mg/mL of each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3, 0.01% bromophenol blue and 33% glycerol

Storage: Upon receipt store at -20°C

Number of Markers 14
Ready to Load Yes
Size Range 10 to 200 kDa
Molecular Weight (g/mol) 200, 150, 120, 100, 85, 70, 60, 50, 40, 30, 25, 20, 15, 10 kDa
Quantity 8 x 250 μL
Product Line PageRuler
Product Type Protein Ladder
Stain Type Unstained
System Type SDS-PAGE

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.