missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PageRuler™ Unstained Low Range Protein Ladder

Product Code. 17126108
Change view
Click to view available options
Quantity:
2 x 250 μL
8 x 250 μL
Unit Size:
Pack of 2
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17126108 8 x 250 μL Pack of 4
11802134 2 x 250 μL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17126108 Supplier Thermo Scientific™ Supplier No. 26632X4
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Thermo Scientific PageRuler Unstained Low Range Protein Ladder is a mixture of eight purified proteins and peptides (3.4 to 100 kDa) for use as size standards in peptide and small-protein gel electrophoresis.

Thermo Scientific™ PageRuler Unstained Low Range Protein Ladder is a mixture of eight (8) purified proteins and peptides (3.4 to 100kDa) for use as size standards in peptide and small protein gel electrophoresis.

Thermo Scientific PageRuler Unstained Low Range Protein Ladder is a mixture of eight purified proteins and peptides (3.4 to 100 kDa) for use as size standards in peptide and small-protein gel electrophoresis. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Product features

  • Reference bands—25 kDa band is stronger than the others for easy orientation
  • Tagged—the ladder proteins (except for the 5 and 3.4 kDa peptides) contains an integral Strep-tag™ II sequence and can be detected on western blots using Strep-Tactin™ conjugates

Applications

  • Accurate protein sizing on SDS-polyacrylamide gels and western blots
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Includes eight vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 100 mM DTT, 1 mM NaN3, 0.01% bromophenol blue and 33% glycerol

Upon receipt, store at -20°C.
Number of Markers 8
Ready to Load Yes
Size Range 3.4 to 100 kDa
Molecular Weight (g/mol) 100, 30, 25, 20, 15, 10, 5, 3.4 kDa
Quantity 8 x 250 μL
Product Line PageRuler
Product Type Protein Ladder
Stain Type Unstained
System Type SDS-PAGE

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.