missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAGE4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554908
This item is not returnable.
View return policy
Description
Immunogen sequence: DSSSSVHDLL VAAMSARVRS RSRGRGDGQE APDVVAFVAP GESQQEEPPT DNQDIEPGQE REGTPPIEER KVEGDCQEMD LEKTRSERGD GSDVKEKTPP NPKHAKTKEA GDGQP Highest antigen sequence identity to the following orthologs: Mouse - 25%, Rat - 27%.
This gene is a member of the GAGE family. The GAGE genes are expressed in a variety of tumors and in some fetal and reproductive tissues. This gene is strongly expressed in prostate and prostate cancer. It is also expressed in other male and female reproductive tissues including testis, fallopian tube, uterus, and placenta, as well as in testicular cancer and uterine cancer. The protein encoded by this gene shares sequence similarity with other GAGE/PAGE proteins, and also belongs to a family of CT (cancer-testis) antigens. The protein may play a role in benign and malignant prostate diseases. A related pseudogene is located on chromosome 7.
Specifications
| PAGE4 | |
| Polyclonal | |
| Unconjugated | |
| PAGE4 | |
| CT16.7; FLJ35184; G antigen family C member 1; GAGE-9; GAGEC1; JM27; JM-27; P antigen family member 4; P antigen family, member 4 (prostate associated); PAGE family member 4; PAGE-1; PAGE4; PAGE-4; prostate-associated gene protein 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9506 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60829 | |
| PAGE4 | |
| Recombinant protein corresponding to Human PAGE4. Recombinant protein control fragment (Product #RP-92151). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction