missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PAGE3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16301495
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16301495 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16301495 Supplier Invitrogen™ Supplier No. PA584539

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GERGDGPNVK GEFLPNLEPV KIPEAGEGQP SV Highest antigen sequence indentity to the following orthologs: Mouse - 38%, Rat - 41%.

PAGE-3 (prostate-associated gene 3 protein), also known as GAGED1 (putative G antigen family Dmember 1), is a 113 amino acid protein that belongsto the GAGE family. The gene that encodes PAGE-3 consists of approximately 6,432 bases andmapsto human chromosome Xp11.21. Chromosome X consists of about 153million base pairs and nearly 1,000 genes. The combination of a X and Y chromosome lead to normal male development while two copies of X lead to normal female development. There are a number ofconditionsrelated to an unusual number and combination ofsexchromosomes being inherited, including Turner's syndrome, Klinefelter'ssyndrome and Triple X syndrome. Color blindness, hemophilia, and Duchennemuscular dystrophy arewellknown X chromosome-linked conditionswhich affectmalesmore frequently as males carry a single X chromosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAGE3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAGE3
Gene Accession No. Q5JUK9
Gene Alias CT16.6; G antigen family D member 1; G antigen, family D, 1; GAGED1; P antigen family member 3; P antigen family, member 3 (prostate associated); PAGE family member 3; PAGE3; PAGE-3; Prostate-associated gene 3 protein
Gene Symbols PAGE3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PAGE3. Recombinant protein control fragment (Product #RP-103991).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139793
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.