missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAGE2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11961-1-AP-150UL
This item is not returnable.
View return policy
Description
PAGE-2 (prostate-associated gene 2 protein), also known as P antigen family member 2B, GAGEC2, CT16.4 or GAGEE2, is a 111 amino acid protein that is expressed primarily in testis and belongs to the GAGE family. PAGE-2B (prostate-associated gene 2B protein), also designated putative G antigen family E member 3, P antigen family member 2B, CT16.5 or GAGEE3, also consists of 111 amino acids and is a member of the GAGE family.Specifications
| PAGE2 | |
| Polyclonal | |
| Unconjugated | |
| PAGE2 | |
| G antigen family E member 2, GAGEC2, GAGEE2, PAGE 2, PAGE2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 203569 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7Z2X7 | |
| PAGE2 | |
| PAGE2 Fusion Protein Ag2566 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction