missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAGE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97691-100ul
This item is not returnable.
View return policy
Description
PAGE1 Polyclonal antibody specifically detects PAGE1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PAGE1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:5000 | |
| CT16.3, G antigen, family B, 1 (prostate associated), GAGE9, GAGE-9AL5, GAGEB1G antigen family B member 1, P antigen family, member 1 (prostate associated), PAGE-1G antigen 9, prostate associated gene 1, Prostate-associated gene 1 protein | |
| Produced in rabbits immunized with E. coli-derived Human PAGE1 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 8712 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction