missing translation for 'onlineSavingsMsg'
Learn More

PAGE1 Antibody, Novus Biologicals™

Product Code. 18378719 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378719 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378719 Supplier Novus Biologicals Supplier No. H00008712B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

PAGE1 Polyclonal antibody specifically detects PAGE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAGE1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.4)
Gene Accession No. NP_003776.2
Gene Alias CT16.3, G antigen, family B, 1 (prostate associated), GAGE9, GAGE-9AL5, GAGEB1G antigen family B member 1, P antigen family, member 1 (prostate associated), PAGE-1G antigen 9, prostate associated gene 1, Prostate-associated gene 1 protein
Host Species Mouse
Immunogen PAGE1 (NP_003776.2, 1 a.a. - 146 a.a.) full-length human protein. MGFLRRLIYRRRPMIYVESSEESSDEQPDEVESPTQSQDSTPAEEREDEGASAAQGQEPEADSQELVQPKTGCELGDGPDTKRVCLRNEEQMKLPAEGPEPEADSQEQVHPKTGCERGDGPDVQELGLPNPEEVKTPEEDEGQSQP
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8712
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.