missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-541
This item is not returnable.
View return policy
Description
PAG1 Polyclonal specifically detects PAG1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| PAG1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.4) with 15mM Sodium Azide | |
| PAG1 | |
| Recombinant intracellular fragment (aa 97-432) of human Cbp (PAG). | |
| Protein A purified | |
| RUO | |
| Primary | |
| The polyclonal antibody recognizes Csk-binding protein (Cbp PAG), a 46 kDa ubiquitously expressed transmembrane adaptor protein present in membrane rafts (glycosphingolipid-enriched microdomains), which however migrates on SDS PAGE gels anomalously as an 80 kDa molecule. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:50, Immunohistochemistry-Paraffin 1:200 | |
| CBPMGC138364, Csk-binding protein, PAGFLJ37858, phosphoprotein associated with glycosphingolipid microdomains 1, phosphoprotein associated with glycosphingolipid-enriched microdomains 1, Transmembrane adapter protein PAG, Transmembrane phosphoprotein Cbp | |
| Rabbit | |
| 46 kDa | |
| 0.1 mg | |
| Cell Biology, Signal Transduction | |
| 55824 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction