missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAFAH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550562
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total PAFAH2 protein.
Has a marked selectivity for phospholipids with short acyl chains at the sn-2 position. May share a common physiologic function with the plasma-type enzyme.
Specifications
| PAFAH2 | |
| Polyclonal | |
| Unconjugated | |
| PAFAH2 | |
| 2310074E22Rik; AI507170; HSD-PLA2; PAFAH2; platelet activating factor acetylhydrolase 2; platelet-activating factor acetylhydrolase 2; platelet-activating factor acetylhydrolase 2 40kDa; platelet-activating factor acetylhydrolase 2, 40kDa; platelet-activating factor acetylhydrolase 2, cytoplasmic; SD-PLA2; serine-dependent phospholipase A2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5051 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2.8 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q99487 | |
| PAFAH2 | |
| Synthetic peptide corresponding to a region derived from internal residues of human platelet-activating factor acetylhydrolase 2, 40kDa. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction