missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAFAH2 Monoclonal Antibody (1A8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524418
This item is not returnable.
View return policy
Description
Peptide Sequence: LMKKICAQHE QSRIITVLGS VHRSQTDFAF VTGNLIGKFF STETRGSLDP YEGQEVMVRA MLAFLQKHLD LKEDYNQWNN LIEGIGPSLT PGAPHHLSSL.
Has a marked selectivity for phospholipids with short acyl chains at the sn-2 position. May share a common physiologic function with the plasma-type enzyme.
Specifications
| PAFAH2 | |
| Monoclonal | |
| Unconjugated | |
| PAFAH2 | |
| 2310074E22Rik; AI507170; HSD-PLA2; PAFAH2; platelet activating factor acetylhydrolase 2; platelet-activating factor acetylhydrolase 2; platelet-activating factor acetylhydrolase 2 40kDa; platelet-activating factor acetylhydrolase 2, 40kDa; platelet-activating factor acetylhydrolase 2, cytoplasmic; SD-PLA2; serine-dependent phospholipase A2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 5051 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA | |
| 1A8 | |
| PBS with no preservative; pH 7.4 | |
| Q99487 | |
| PAFAH2 | |
| PAFAH2 (NP_000428, 293 a.a. ~ 392 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction