missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAFAH1B3 Antibody (OTI5D2), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73229AF700
This item is not returnable.
View return policy
Description
PAFAH1B3 Monoclonal specifically detects PAFAH1B3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PAFAH1B3 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PAFAH1B3 | |
| Full length recombinant protein of human PAFAH1B3 (NP_001139411) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI5D2 | |
| Western Blot | |
| EC 3.1.1.47, FLJ44990, PAF acetylhydrolase 29 kDa subunit, PAF-AH 29 kDa subunit, PAFAH subunit gamma, PAF-AH subunit gamma, PAF-AH1b alpha 1 subunit, PAFAHG, platelet-activating factor acetylhydrolase 1b, catalytic subunit 3 (29kDa), platelet-activating factor acetylhydrolase IB subunit gamma, platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit (29kD), platelet-activating factor acetylhydrolase, isoform Ib, gamma subunit 29kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 3 (29kDa) | |
| Mouse | |
| Affinity Purified | |
| Neuroscience | |
| 5050.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction