missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAFAH1B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13725
This item is not returnable.
View return policy
Description
PAFAH1B2 Polyclonal specifically detects PAFAH1B2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PAFAH1B2 | |
| Polyclonal | |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| EC 3.1.1.47, intracellular platelet-activating factor acetylhydrolase alpha 2 subunit, PAF acetylhydrolase 30 kDa subunit, PAF-AH 30 kDa subunit, PAFAH subunit beta, PAF-AH subunit beta, PAF-AH1b alpha 2 subunit, PAFAHB, platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa), platelet-activating factor acetylhydrolase IB subunit beta, platelet-activating factor acetylhydrolase, isoform Ib, beta subunit 30kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5049 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PAFAH1B2 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: QLINTRQPQAKIIVLGLLPRGEKPNPLRQKNAKVNQLLKVSLPKLANVQLLDTDG | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction