missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAFAH1B2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93856-0.02ml
This item is not returnable.
View return policy
Description
PAFAH1B2 Polyclonal antibody specifically detects PAFAH1B2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PAFAH1B2 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| EC 3.1.1.47, intracellular platelet-activating factor acetylhydrolase alpha 2 subunit, PAF acetylhydrolase 30 kDa subunit, PAF-AH 30 kDa subunit, PAFAH subunit beta, PAF-AH subunit beta, PAF-AH1b alpha 2 subunit, PAFAHB, platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa), platelet-activating factor acetylhydrolase IB subunit beta, platelet-activating factor acetylhydrolase, isoform Ib, beta subunit 30kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-229 of human PAFAH1B2 (NP_002563.1). MSQGDSNPAAIPHAAEDIQGDDRWMSQHNRFVLDCKDKEPDVLFVGDSMVQLMQQYEIWRELFSPLHALNFGIGGDTTRHVLWRLKNGELENIKPKVIVVWVGTNNHENTAEEVAGGIEAIVQLINTRQPQAKIIVLGLLPRGEKPNPLRQKNAKVNQLLKVSLPKLANVQLLDTDGGFVHSDGAISCHDMFDFLHLTGGGYAKICKPLHELIMQLLEETPEEKQTTIA | |
| 0.02 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 5049 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction