Learn More
PAFAH Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29304-1-AP-150UL
This item is not returnable.
View return policy
Description
The PAFAH antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human PAFAH. This antibody recognizes human, mouse, rat antigen. The PAFAH antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| PAFAH | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| 150 μL | |
| 7941 | |
| IgG |