missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAF49 Polyclonal Antibody

PAF49 Polyclonal Antibody
Brand: Thermo Scientific PA5114981
This item is not returnable.
View return policy
Description
PAF49 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| PAF49 | |
| Polyclonal | |
| Unconjugated | |
| Cd3eap | |
| 1190028F09; 2610103M17Rik; A34.5; antis; Anti-sense to ERCC-1 protein; antisense to ERCC-1 protein; Ase1; ASE-1; CAST; CD3 epsilon associated protein; antisense to Ercc1; CD3E antigen, epsilon polypeptide associated protein; CD3E antigen, epsilon polypeptide-associated protein; CD3e molecule associated protein; CD3e molecule, epsilon associated protein; Cd3eap; CD3E-associated protein; CD3-epsilon-associated protein; DNA-directed RNA polymerase I subunit RPA34; PAF49; RGD1305916; RNA polymerase I-associated factor PAF49 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10849 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O15446 | |
| Cd3eap | |
| A synthesized peptide derived from human CD3EAP(Accession O15446), corresponding to amino acid residues L441-G491. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction