missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAF1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15441-1-AP-20UL
This item is not returnable.
View return policy
Description
The MW of this protein is 80 kDa, and this antibody specifically recognizes the 80 kDa protein.
This gene encodes a subunit of the polymerase associated factor (PAF1) complex. The PAF1 complex interacts with RNA polymerase II and plays a role in transcription elongation as well as histone modifications including ubiquitylation and methylation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
| PAF1 | |
| Polyclonal | |
| Unconjugated | |
| PAF1 | |
| F23149_1, hPAF1, PAF1, PD2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54623 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N7H5 | |
| PAF1 | |
| PAF1 Fusion Protein Ag7212 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction