missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115713
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PAF1.
This gene encodes a subunit of the polymerase associated factor (PAF1) complex. The PAF1 complex interacts with RNA polymerase II and plays a role in transcription elongation as well as histone modifications including ubiquitylation and methylation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| PAF1 | |
| Polyclonal | |
| Unconjugated | |
| Paf1 | |
| 5730511K23Rik; AA675045; F23149_1; hPAF1; Paf1; PAF1 homolog, Paf1/RNA polymerase II complex component; Paf1, RNA polymerase II associated factor, homolog; Paf1, RNA polymerase II associated factor, homolog (S. cerevisiae); pancreatic differentiation protein 2; PD2; RGD1306219; RNA polymerase II-associated factor 1 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 361531, 54623, 54624 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q4V886, Q8K2T8, Q8N7H5 | |
| Paf1 | |
| A synthesized peptide derived from human PAF1(Accession Q8N7H5), corresponding to amino acid residues M1-L39. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction