missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAEP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554152
This item is not returnable.
View return policy
Description
Immunogen sequence: NYTVANEATL LDTDYDNFLF LCLQDTTTPI QSMMCQYLAR VLVEDDEIMQ GFIRAFRPLP RHLWYLLDLK QME Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 26%.
Human placental protein 14 (PP14; also known as glycodelin and progesterone-associated endometrial protein) is a protein of the lipocalin structural superfamily. PP14 is the most abundant product of the secretory endometrium, and has been proposed as a biochemical marker of endometrial function in women undergoing in vitro fertilization treatment.
Specifications
| PAEP | |
| Polyclonal | |
| Unconjugated | |
| PAEP | |
| alpha uterine protein; GD; GdA; GdF; GdS; Glycodelin; glycodelin-A; glycodelin-F; glycodelin-S; PAEG; PAEP; PEG; PEP; placental protein 14; PP14; PP14 protein (placental protein 14); pregnancy-associated endometrial alpha-2 globulin; pregnancy-associated endometrial alpha-2-globulin; progestagen associated endometrial protein; Progestagen-associated endometrial protein; progestagen-associated endometrial protein (placental protein 14, pregnancy-associated endometrial a; progesterone-associated endometrial protein; ZIF-1; Zona-binding inhibitory factor-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5047 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P09466 | |
| PAEP | |
| Recombinant protein corresponding to Human PAEP. Recombinant protein control fragment (Product #RP-92930). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur